missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Ryk Antibody (F35 P7 D7 F5), Novus Biologicals™

Mouse Monoclonal Antibody

€ 213.00 - € 508.00

Specifications

Antigen Ryk
Clone F35 P7 D7 F5
Dilution Western Blot : 1-5 μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067251
View Documents
Novus Biologicals™
NBP3-30896-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30066879
View Documents
Novus Biologicals™
NBP3-30896-200ug
200 μg
€ 508.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ryk Monoclonal antibody specifically detects Ryk in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

Ryk
Western Blot : 1-5 μg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
D3S3195, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, JTK5, JTK5A, JTK5A protein tyrosine kinase, RYK receptor-like tyrosine kinase, RYK1, tyrosine-protein kinase RYK
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human Ryk protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse and rat.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
F35 P7 D7 F5
Western Blot
Unconjugated
Mouse
Protein Kinase
Phosphate buffered saline
6259
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.