Learn More
(S)-(+)-1-Boc-3-bromopyrrolidine, 95%
Product Code. 15443878
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443878 250 mg 250mg
15433878 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15443878 Supplier Thermo Scientific Alfa Aesar Supplier No. H57408.MD
Request Bulk or Custom Format
CAS: 569660-89-5 | C9H16BrNO2 | 250.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 569660-89-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD17214728
InChI Key QJTKPXFJOXKUEY-UHFFFAOYNA-N
Synonym s-tert-butyl 3-bromopyrrolidine-1-carboxylate, s-3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3s-3-bromopyrrolidine-1-carboxylate, s-+-1-boc-3-bromopyrrolidine, tert-butyl s-3-bromopyrrolidine-1-carboxylate, s-3-bromopyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 34179480
IUPAC Name tert-butyl (3S)-3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Br)C1
Specifications
Quantity 250 mg
UN Number UN3077
Optical Rotation +33° (c=0.8 in Chloroform)
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (S)-(+)-1-Boc-3-bromopyrrolidine