Learn More
S-(2,4-Dinitrophenyl)-Glutathione, TRC
Specifications
Specifications
| Chemical Name or Material | S-(2,4-Dinitrophenyl)-Glutathione |
| CAS | 26289-39-4 |
| Molecular Formula | C16 H19 N5 O10 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C16H19N5O10S/c17-9(16(26)27)2-4-13(22)19-10(15(25)18-6-14(23)24)7-32-12-3-1-8(20(28)29)5-11(12)21(30)31/h1,3,5,9-10H,2,4,6-7,17H2,(H,18,25)(H,19,22)(H,23,24)(H,26,27)/t9-,10-/m0/s1 |
| SMILES | N[C@@H](CCC(=O)N[C@@H](CSc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)NCC(=O)O)C(=O)O |
| IUPAC Name | (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-(2,4-dinitrophenyl)sulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid |
| Molecular Weight (g/mol) | 473.41 |
| Formula Weight | 473.0853 |