Learn More
(S)-2-Aminohexanedioic Acid 6-Methyl Ester Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-2-Aminohexanedioic Acid 6-Methyl Ester Hydrochloride |
| CAS | 147780-39-0 |
| Molecular Formula | C7H14ClNO4 |
| Quantity | 5 g |
| Synonym | H-Homoglu(ome)-OH-HCl, H-Aad(OMe)-OH.HC, (S)-2-aminoheptanedioic acid |
| InChI Formula | InChI=1S/C7H13NO4.ClH/c1-12-6(9)4-2-3-5(8)7(10)11;/h5H,2-4,8H2,1H3,(H,10,11);1H/t5-;/m0./s1 |
| SMILES | Cl.COC(=O)CCCC(N)C(=O)O |
| IUPAC Name | (2S)-2-amino-6-methoxy-6-oxohexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 211.64 |
| Formula Weight | 211.0611 |