Learn More
(S)-2-Aminohexanedioic Acid 6-Methyl Ester Hydrochloride, TRC
Product Code. 30313647
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313647 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30313647 Supplier Toronto Research Chemicals Supplier No. TRCA610010250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-2-Aminohexanedioic Acid 6-Methyl Ester Hydrochloride
CAS 147780-39-0
Molecular Formula C7H14ClNO4
Quantity 250 mg
Synonym H-Homoglu(ome)-OH-HCl, H-Aad(OMe)-OH.HC, (S)-2-aminoheptanedioic acid
InChI Formula InChI=1S/C7H13NO4.ClH/c1-12-6(9)4-2-3-5(8)7(10)11;/h5H,2-4,8H2,1H3,(H,10,11);1H/t5-;/m0./s1
SMILES Cl.COC(=O)CCCC(N)C(=O)O
IUPAC Name (2S)-2-amino-6-methoxy-6-oxohexanoic acid;hydrochloride
Molecular Weight (g/mol) 211.64
Formula Weight 211.0611