Learn More
(S)-2-Aminohexanedioic Acid 6-Methyl Ester Hydrochloride, TRC
Product Code. 30326930
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326930 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326930 Supplier Toronto Research Chemicals Supplier No. TRCA6100101G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-2-Aminohexanedioic Acid 6-Methyl Ester Hydrochloride
CAS 147780-39-0
Molecular Formula C7H14ClNO4
Quantity 1 g
Synonym H-Homoglu(ome)-OH-HCl, H-Aad(OMe)-OH.HC, (S)-2-aminoheptanedioic acid
InChI Formula InChI=1S/C7H13NO4.ClH/c1-12-6(9)4-2-3-5(8)7(10)11;/h5H,2-4,8H2,1H3,(H,10,11);1H/t5-;/m0./s1
SMILES Cl.COC(=O)CCCC(N)C(=O)O
IUPAC Name (2S)-2-amino-6-methoxy-6-oxohexanoic acid;hydrochloride
Molecular Weight (g/mol) 211.64
Formula Weight 211.0611