missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Benzyloxycarbonylamino)butyric acid, 98%, Thermo Scientific Chemicals

Product Code. 15478928
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15478928

Brand: Thermo Scientific Alfa Aesar H62426.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42918-86-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00077006
InChI Key SZQMTCSQWUYUML-JTQLQIEISA-N
Synonym z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid
PubChem CID 7349998
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 237.25
Percent Purity 98%
Chemical Name or Material (S)-2-(Benzyloxycarbonylamino)butyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.