missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%

Product Code. p-7034295
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15438427

Brand: Thermo Scientific Alfa Aesar H51966.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100564-78-1 | C15H21NO4 | 279.336 g/mol

(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100564-78-1
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00076904
InChI Key MCODLPJUFHPVQP-LBPRGKRZSA-N
Synonym boc-l-homophenylalanine, boc-homophe-oh, boc-hophe-oh, boc-l-homo-phe, s-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid, boc-homophenylalanine, boc-hfe-oh, n-boc-l-homophenylalanine, n-alpha-boc-l-homophenylalanine
PubChem CID 7018726
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 76°C to 80°C
Beilstein 3653506
Solubility Information Soluble in methanol.
Quantity 1 g
Formula Weight 279.34
Optical Rotation +6.5° (c=2 in Ethanol)
Percent Purity 98%
Chemical Name or Material (S)-2-(Boc-amino)-4-phenylbutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.