Learn More
S-(2-Carboxyethyl)-L-cysteine, TRC
Product Code. 30349423
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349423 50 mg 50mg
30368343 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30349423 Supplier Toronto Research Chemicals Supplier No. TRCC17805550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100429-59-2
Molecular Formula C6H11NO4S
Molecular Weight (g/mol) 193.22
Synonym L-3-[(2-Carboxyethyl)thio]alanine, 2-Amino-4-thiaheptanedioic Acid, S-(2-Carboxyethyl)cysteine, CEC:
IUPAC Name (2S)-2-amino-3-(2-carboxyethylsulfanyl)propanoic acid
SMILES N[C@H](CSCCC(=O)O)C(=O)O
Specifications
InChI Formula InChI=1S/C6H11NO4S/c7-4(6(10)11)3-12-2-1-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)/t4-/m1/s1
Quantity 50 mg
Formula Weight 193.0409
Shipping Condition Room Temperature
Chemical Name or Material S-(2-Carboxyethyl)-L-cysteine