missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Chloromandelic acid, 95%, Thermo Scientific Chemicals

Catalog Number 15447658
20.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52950-19-3
C8H7ClO3
186.591
MFCD00798437
RWOLDZZTBNYTMS-ZETCQYMHSA-N
s-+-2-chloromandelic acid, s-2-2-chlorophenyl-2-hydroxyacetic acid, s-2-chloromandelic acid, s-o-chloromandelicacid, 2-chloro-l-mandelic acid, 2s-2-2-chlorophenyl-2-hydroxy-acetic acid, s-2-chlorophenyl hydroxy acetic acid, pubchem5743, s-o-chloromandelic acid, ksc914s2j
6922874
(2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
C1=CC=C(C(=C1)C(C(=O)O)O)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52950-19-3
C8H7ClO3
186.591
MFCD00798437
RWOLDZZTBNYTMS-ZETCQYMHSA-N
s-+-2-chloromandelic acid, s-2-2-chlorophenyl-2-hydroxyacetic acid, s-2-chloromandelic acid, s-o-chloromandelicacid, 2-chloro-l-mandelic acid, 2s-2-2-chlorophenyl-2-hydroxy-acetic acid, s-2-chlorophenyl hydroxy acetic acid, pubchem5743, s-o-chloromandelic acid, ksc914s2j
6922874
(2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
C1=CC=C(C(=C1)C(C(=O)O)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 122°C
Color Brown to White
Quantity 1 g
Physical Form Powder
Formula Weight 186.59
Percent Purity 95%
Chemical Name or Material (S)-(+)-2-Chloromandelic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'