missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methylbutyric Acid, TRC

Product Code. 30479963
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479963 1 g 1g
30483061 500 mg 500mg
30475312 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479963 Supplier Toronto Research Chemicals Supplier No. TRCM2943551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1730-91-2
Molecular Formula C5 H10 O2
Molecular Weight (g/mol) 102.132
Synonym (2S)-2-Methyl-butanoic Acid, (+)-2-Methylbutanoic Acid, (+)-2-Methylbutyric Acid, (+)-alpha-Methylbutyric Acid, (2S)-2-Methylbutanoic Acid, (2S)-2-Methylbutyric Acid, (S)-2-Methylbutanoic Acid, (S)-2-Methylbutyric Acid, S-(+)-2-Methylbutanoic Acid
IUPAC Name (2S)-2-methylbutanoic acid
SMILES CC[C@H](C)C(=O)O

Tekniske data

Quantity 1 g
InChI Formula InChI=1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/t4-/m0/s1
Formula Weight 102.068
Chemical Name or Material (S)-(+)-2-Methylbutyric Acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.