Learn More
(S)-3,4-Dihydro-5,12-dihydroxy-7-methoxy-4-[[2,3,6-trideoxy-3-[(trifluoroacetyl)amino]-α-L-lyxo-hexopyranosyl]oxy]-2,6,11(1H)-naphthacenetrione (>75% by HPLC), TRC
Specifications
Specifications
| Chemical Name or Material | (S)-3,4-Dihydro-5,12-dihydroxy-7-methoxy-4-((2,3,6-trideoxy-3-((trifluoroacetyl)amino)-alpha-L-lyxo-hexopyranosyl)oxy)-2,6,11(1H)-naphthacenetrione |
| CAS | 64882-10-6 |
| Molecular Formula | C27H24F3NO10 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C27H24F3NO10/c1-9-21(33)13(31-26(38)27(28,29)30)8-16(40-9)41-15-7-10(32)6-12-18(15)25(37)20-19(23(12)35)22(34)11-4-3-5-14(39-2)17(11)24(20)36/h3-5,9,13,15-16,21,33,35,37H,6-8H2,1-2H3,(H,31,38) |
| SMILES | O=C(C1)C[C@H](O[C@H]2C[C@@H](NC(C(F)(F)F)=O)[C@@H](O)[C@H](C)O2)C(C1=C3O)=C(O)C4=C3C(C5=CC=CC(OC)=C5C4=O)=O |
| Molecular Weight (g/mol) | 579.48 |