Learn More
(S)-5-Amino-4-(4-amino-1-oxoisoindolin-2-yl)-5-oxopentanoic Acid, TRC
Specifications
Specifications
| CAS | 874760-72-2 |
| Analyte or Component Names | (S)-5-Amino-4-(4-amino-1-oxoisoindolin-2-yl)-5-oxopentanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 N3 O4 |
| Synonym | (γS)-4-Amino-γ-(aminocarbonyl)-1,3-dihydro-1-oxo-2H-isoindole-2-butanoic Acid, 2H-Isoindole-2-butanoic acid, 4-amino-γ-(aminocarbonyl)-1,3-dihydro-1-oxo-, (γS)- (9CI, ACI), (γS)-4-Amino-γ-(aminocarbonyl)-1,3-dihydro-1-oxo-2H-isoindole-2-butanoic acid (ACI), 2H-Isoindole-2-butanoic acid 4-amino-γ-(aminocarbonyl)-1,3-dihydro-1-oxo-, (γS)- |
| InChI Formula | InChI=1S/C13H15N3O4/c14-9-3-1-2-7-8(9)6-16(13(7)20)10(12(15)19)4-5-11(17)18/h1-3,10H,4-6,14H2,(H2,15,19)(H,17,18)/t10-/m0/s1 |
| Solution Type | Neat |
| SMILES | NC(=O)[C@H](CCC(=O)O)N1Cc2c(N)cccc2C1=O |
| IUPAC Name | (4S)-5-amino-4-(4-amino-1-oxo-isoindolin-2-yl)-5-oxo-pentanoic acid |
| Industry Type | Pharmaceutical |
| Show More |