missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S-Adenosyl-Homocysteine Monoclonal Antibody (3B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549612
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
S-Adenosylhomocysteine (AdoHcy) is the immediate precursor of all of the homocysteine produced in the body. The reaction is catalyzed by S-adenosylhomocysteine hydrolase and is reversible with the equilibrium favoring formation of AdoHcy. In vivo, the reaction is driven in the direction of homocysteine formation by the action of the enzyme adenosine deaminase, which converts the second product of the S-adenosylhomocysteine hydrolase reaction, adenosine, to inosine. Except for methyl transfer from betaine and from methylcobalamin in the methionine synthase reaction, AdoHcy is the product of all methylation reactions that involve S-adenosylmethionine (AdoMet) as the methyl donor. Methylation is significant in epigenetic regulation of protein expression via DNA and histone methylation. The inhibition of these AdoMet-mediated processes by AdoHcy is a proven mechanism for metabolic alteration. Because the conversion of AdoHcy to homocysteine is reversible, with the equilibrium favoring the formation of AdoHcy, increases in plasma homocysteine are accompanied by an elevation of AdoHcy in most cases. Disturbances in the transmethylation pathway indicated by abnormal S-adenosylmethionine, S-adenosylhomocysteine or their ratio have been reported in many neurodegenerative diseases, such as dementia, depression or Parkinson's disease. (PMID: 18065573, 17892439). [National Center for Biotechnology Information. PubChem Compound Database; CID=439155].
Specifications
| S-Adenosyl-Homocysteine | |
| Monoclonal | |
| 5.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 3B3 | |
| Unconjugated | |
| AdoHcy; C14H20N6O5S; Formycinylhomocysteine; S-(5'-adenosyl)-L-homocysteine; S-adenosylhomocysteine; S-adenosyl-L-homocysteine | |
| Conjugate of SAH hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction