missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-arrestin Antibody (PDS-1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2385AF350
This item is not returnable.
View return policy
Description
S-arrestin Monoclonal specifically detects S-arrestin in Human, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| S-arrestin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SAG | |
| Porcine retinal S-arrestin [Swiss-ProtNo. P79260] | |
| 0.1 mL | |
| Primary | |
| Human, Pig, Bovine | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PDS-1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin | |
| Mouse | |
| Protein G purified | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 6295.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction