Learn More
(S)-Cinacalcet Hydrochloride, Mikromol™
Description
(S)-Cinacalcet Hydrochloride
Specifications
Specifications
| Chemical Name or Material | ent-Cinacalcet Hydrochloride |
| CAS | 1217809-88-5 |
| Analyte or Component Names | ent-Cinacalcet Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H22 F3 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Naphthalenemethanamine, α-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride (1:1), (αS)-, (S)-Cinacalcet hydrochloride, ent-Cinacalcet hydrochloride, Cinacalcet Related Compound D as Hydrochloride |
| InChI Formula | InChI=1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/t16-;/m0./s1 |
| SMILES | Cl.C[C@H](NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23 |
| IUPAC Name | N-[(1S)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine;hydrochloride |
| Show More |