Learn More
(S)-cis-Verbenol, 97%, sum of isomers
Product Code. p-3779877 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. 10224634
Brand: Thermo Scientific Acros 347330050
Request Bulk or Custom Format
CAS: 18881-04-4 | C10H16O | 152.24 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18881-04-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00065444
InChI Key WONIGEXYPVIKFS-YIZRAAEISA-N
Synonym s-cis-verbenol, verbenol, s-cis, unii-xr9t57f48t, cis-verbenol, bicyclo 3.1.1 hept-3-en-2-ol, 4,6,6-trimethyl-, 1s,2s,5s, 1s-cis-verbenol, ccris 5289, ?-cis-verbenol
PubChem CID 87839
IUPAC Name (1S,2S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
SMILES CC1=CC(C2CC1C2(C)C)O
Specifications
CAS Min % 96.0
CAS Max % 100.0
Melting Point 62°C to 68°C
Color Bordeaux to Brown-Red
Density 85g/mL
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (sum of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.49
Quantity 5 g
Specific Rotation Condition + 10.00 (20.00°C c=2,C2H5OH)
Specific Rotation + 10
Solubility Information Solubility in water: insoluble
Formula Weight 152.24
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-cis-Verbenol, Sum of isomers