Learn More
(S)-(-)-Dropropizine, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-(-)-Dropropizine |
| CAS | 99291-25-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13H20N2O2 |
| Quantity | 50 mg |
| Synonym | Levodropropizine, (2S)-3-(4-Phenylpiperazin-1-yl)propane-1,2-diol |
| InChI Formula | InChI=1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m0/s1 |
| SMILES | OC[C@@H](O)CN1CCN(CC1)c2ccccc2 |
| Molecular Weight (g/mol) | 236.31 |
| Formula Weight | 236.1525 |
| Show More |