Learn More
(S) FTY720 Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | (S) FTY720 Phosphate |
| CAS | 402616-26-6 |
| Molecular Formula | C19 H34 N O5 P |
| Quantity | 10 mg |
| Synonym | 1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, 1-(dihydrogen phosphate), (2S)-, 1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, mono(dihydrogen phosphate) (ester), (2S)- (9CI), (+)-(S)-FTY 720 phosphate, (S)-FTY 720P, (S)-Fingolimod O-phosphate |
| InChI Formula | InChI=1S/C19H34NO5P/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-25-26(22,23)24/h9-12,21H,2-8,13-16,20H2,1H3,(H2,22,23,24)/t19-/m0/s1 |
| SMILES | CCCCCCCCc1ccc(CC[C@](N)(CO)COP(=O)(O)O)cc1 |
| IUPAC Name | [(2S)-2-amino-2-(hydroxymethyl)-4-(4-octylphenyl)butyl] dihydrogen phosphate |
| Molecular Weight (g/mol) | 387.45 |
| Formula Weight | 387.2175 |