Learn More
(S)-GNE-140, MedChemExpress
Product Code. 16493148
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16493148
Brand: Medchem Express HY100742B
MedChemExpress (S)-GNE-140 is the less active enantiomer of GNE-140 which can inhibit Lactate dehydrogenase A (LDHA).
(S)-GNE-140 is the less active enantiomer of GNE-140 which can inhibit Lactate dehydrogenase A (LDHA).
Specifications
Chemical Name or Material (S)-GNE-140
Physical Form Solid
CAS 2003234-64-6
Color Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H23ClN2O3S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 14 mg/mL (28.05 mM; Need ultrasonic and warming)
SMILES O=C1N[C@](C2=CSC=C2)(C3=CC=C(N4CCOCC4)C=C3)CC(O)=C1SC5=CC=CC=C5Cl
Molecular Weight (g/mol) 499.04
Formula Weight 499.04
Percent Purity 99.04%
Grade Research
Show More Show Less
Research purposes only