missing translation for 'onlineSavingsMsg'
Learn More

(S)-Ibuprofen (S)-Methocarbamol Ester, TRC

Product Code. 30404639
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404639 10 mg 10mg
30398940 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404639 Supplier Toronto Research Chemicals Supplier No. TRCI14007010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Ibuprofen (S)-Methocarbamol Ester
Analyte or Component Names (S)-Ibuprofen (S)-Methocarbamol Ester
Shipping Condition Room Temperature
Molecular Formula C24 H31 N O6
InChI Formula InChI=1S/C24H31NO6/c1-16(2)13-18-9-11-19(12-10-18)17(3)23(26)31-20(15-30-24(25)27)14-29-22-8-6-5-7-21(22)28-4/h5-12,16-17,20H,13-15H2,1-4H3,(H2,25,27)/t17-,20-/m0/s1
Solution Type Neat
SMILES COc1ccccc1OC[C@@H](COC(=O)N)OC(=O)[C@@H](C)c2ccc(CC(C)C)cc2
IUPAC Name [(2S)-1-carbamoyloxy-3-(2-methoxyphenoxy)propan-2-yl] (2S)-2-[4-(2-methylpropyl)phenyl]propanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 429.506
Quantity 10 mg
Formula Weight 429.215
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.