missing translation for 'onlineSavingsMsg'
Learn More

(S)-Lisinopril Dihydrate, TRC

Product Code. 30137876
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137876 250 mg 250mg
30137884 1 g 1g
18033120 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137876 Supplier Toronto Research Chemicals Supplier No. TRCL468985250MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83915-83-7
Synonym Lisinopril Dihydrate∣N2-[(1S)-1-Carboxy-3-phenylpropyl]-L-lysyl-L-proline dihydrate∣(S)-1-[N2-(1-Carboxy-3-phenylpropyl)-L-lysyl]-L-proline dihydrate

Specifications

Format Neat
Molecular Weight (g/mol) 441.52
Product Type Metabolite
Quantity 250 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.