Learn More
(S)-Lisinopril Dihydrate, TRC
Product Code. 30137884
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137884 1 g 1g
30137876 250 mg 250mg
18033120 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137884 Supplier Toronto Research Chemicals Supplier No. TRCL4689851G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83915-83-7
Synonym Lisinopril Dihydrate∣N2-[(1S)-1-Carboxy-3-phenylpropyl]-L-lysyl-L-proline dihydrate∣(S)-1-[N2-(1-Carboxy-3-phenylpropyl)-L-lysyl]-L-proline dihydrate
Specifications
Format Neat
Molecular Weight (g/mol) 441.52
Product Type Metabolite
Quantity 1 g
Shipping Condition Room Temperature