Learn More
(S)-methyl 5-Oxo-5,6,7,8-tetrahydroimidazo[1,5-c]pyrimidine-7-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-methyl 5-Oxo-5,6,7,8-tetrahydroimidazo[1,5-c]pyrimidine-7-carboxylate |
| CAS | 69614-04-6 |
| Molecular Formula | C8H9N3O3 |
| Quantity | 5 g |
| Synonym | (7S)-5,6,7,8-Tetrahydro-5-oxo-Imidazo[1,5-c]pyrimidine-7-carboxylic Acid Methyl Ester |
| InChI Formula | InChI=1S/C8H9N3O3/c1-14-7(12)6-2-5-3-9-4-11(5)8(13)10-6/h3-4,6H,2H2,1H3,(H,10,13)/t6-/m0/s1 |
| SMILES | O=C([C@@H](N1)CC2=CN=CN2C1=O)OC |
| IUPAC Name | methyl (7S)-5-oxo-7,8-dihydro-6H-imidazo[1,5-c]pyrimidine-7-carboxylate |
| Molecular Weight (g/mol) | 195.18 |
| Formula Weight | 195.0644 |