Learn More
S-Methyl Tiopronin, TRC
Specifications
Specifications
| Chemical Name or Material | S-Methyl Tiopronin |
| CAS | 87254-91-9 |
| Molecular Formula | C6 H11 N O3 S |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C6H11NO3S/c1-4(11-2)6(10)7-3-5(8)9/h4H,3H2,1-2H3,(H,7,10)(H,8,9) |
| SMILES | CSC(C)C(=O)NCC(=O)O |
| IUPAC Name | 2-(2-methylsulfanylpropanoylamino)acetic acid |
| Molecular Weight (g/mol) | 177.22 |
| Formula Weight | 177.046 |