missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N α-Dimethylbenzylamine, 99+%, Thermo Scientific Chemicals

Catalog Number 11905251
111.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19131-99-8
C9H13N
135.21
RCSSHZGQHHEHPZ-QMMMGPOBSA-N
s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
2060073
(1S)-N-methyl-1-phenylethanamine
CC(C1=CC=CC=C1)NC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19131-99-8
C9H13N
135.21
RCSSHZGQHHEHPZ-QMMMGPOBSA-N
s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
2060073
(1S)-N-methyl-1-phenylethanamine
CC(C1=CC=CC=C1)NC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9240g/mL
Boiling Point 74.0°C to 76.0°C (11.0 mmHg)
Flash Point 61°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NHCH3
Quantity 5 g
Specific Gravity 0.924
Solubility Information Solubility in water: insoluble
Formula Weight 135.21
Percent Purity 99+%
Chemical Name or Material (S)-(-)-N α-Dimethylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-N alpha-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'