missing translation for 'onlineSavingsMsg'
Learn More

(S)-Naproxen Methyl Ester, TRC

Product Code. 30465864
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465864 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465864 Supplier Toronto Research Chemicals Supplier No. TRCN377550500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate (Naproxen Methyl Ester)
CAS 26159-35-3
Purity Grade Notes HPLC
Molecular Formula C15 H16 O3
Quantity 500 mg
Synonym Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate, Naproxen Imp. E (EP), Naproxen Methyl Ester, 2-Naphthaleneacetic acid','6-methoxy-α-methyl-','methyl ester','(αS)- (9CI), 2-Naphthaleneacetic acid','6-methoxy-α-methyl-','methyl ester','(S)- (8CI), (+)-Naproxen methyl ester, (S)-Naproxen methyl ester, Naproxen methyl ester
InChI Formula InChI=1S/C15H16O3/c1-10(15(16)18-3)11-4-5-13-9-14(17-2)7-6-12(13)8-11/h4-10H,1-3H3/t10-/m0/s1
SMILES COC(=O)[C@@H](C)c1ccc2cc(OC)ccc2c1
IUPAC Name methyl (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
Molecular Weight (g/mol) 244.29
Formula Weight 244.1099
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.