Learn More
S-Phenyl-d5-mercapturic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Mercapturic acid S-phenyl-d5 |
| CAS | 2519846-58-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C11 2H5 H8 N O3 S |
| Quantity | 10 mg |
| Synonym | N-Acetyl-S-phenyl-d5-L-cysteine |
| InChI Formula | InChI=1S/C11H13NO3S/c1-8(13)12-10(11(14)15)7-16-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3,(H,12,13)(H,14,15)/t10-/m0/s1/i2D,3D,4D,5D,6D |
| SMILES | [2H]c1c([2H])c([2H])c(SC[C@H](NC(=O)C)C(=O)O)c([2H])c1[2H] |
| IUPAC Name | (2R)-2-acetamido-3-(2,3,4,5,6-pentadeuteriophenyl)sulfanylpropanoic acid |
| Molecular Weight (g/mol) | 244.32 |
| Show More |