missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S Protein Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF814495
This item is not returnable.
View return policy
Description
S Protein Monoclonal antibody specifically detects S Protein in Virus samples. It is validated for ELISASpecifications
| S Protein | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Lyophilized |
| ELISA | |
| OTI2D2 | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli. | |
| 100 μg | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction