missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress (S,R,S)-AHPC-C6-PEG3-C4-Cl
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
The connector of linker is Halogen group. (S,R,S)-AHPC-C6-PEG3-C4-Cl incorporates the (S,R,S)-AHPC based VHL ligand and an alkyl/ether-based linker. (S,R,S)-AHPC-C6-PEG3-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.
Specifications
Specifications
| Molecular Weight (g/mol) | 751.42 g/mol |
| CAS | 1835705-55-9 |
| Content And Storage | Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 500 mg |
| Synonym | VH032-C6-PEG3-C4-Cl ∣ VHL Ligand-Linker Conjugates 12 ∣ E3 ligase Ligand-Linker Conjugates 8 |
| Solubility | DMSO : ≥ 100 mg/mL (133.08 mM) ∣ Ethanol : 100 mg/mL (133.08 mM; Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction