Learn More
MedChemExpress (S,R,S)-AHPC-PEG4-NH2
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Specifications
Specifications
| Molecular Weight (g/mol) | 663.83 g/mol |
| CAS | 2010159-57-4 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 100 mg |
| Synonym | VH032-PEG4-NH2 ∣ VHL Ligand-Linker Conjugates 4 ∣ E3 ligase Ligand-Linker Conjugates 7 Free Base |
| Solubility | DMSO : 170 mg/mL (256.09 mM; Need ultrasonic) |