missing translation for 'onlineSavingsMsg'
Learn More

(S)-Ropivacaine N-Isopropyl, TRC

Product Code. 30414840
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414840 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414840 Supplier Toronto Research Chemicals Supplier No. TRCR6749901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Ropivacaine N-Isopropyl
CAS 265120-58-9
Purity Grade Notes HPLC
Molecular Formula C17H26N2O
Quantity 1 g
Synonym (2S)-N-(2,6-Dimethylphenyl)-1-(1-methylethyl)-2-piperidinecarboxamide, Ropivacaine EP Impurity E
InChI Formula InChI=1S/C17H26N2O/c1-12(2)19-11-6-5-10-15(19)17(20)18-16-13(3)8-7-9-14(16)4/h7-9,12,15H,5-6,10-11H2,1-4H3,(H,18,20)/t15-/m0/s1
SMILES O=C([C@@H]1N(C(C)C)CCCC1)NC2=C(C)C=CC=C2C
IUPAC Name (2S)-N-(2,6-dimethylphenyl)-1-propan-2-ylpiperidine-2-carboxamide
Molecular Weight (g/mol) 274.4
Formula Weight 274.2045
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.