Learn More
(S,S)-Metalaxyl Metabolite CGA 226048, Honeywell Fluka™
Product Code. 15949771 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15949771 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15949771 Supplier Honeywell Fluka Supplier No. R152210MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
Physical Form Solid
Industry Type Fungicides and Bactericides
Quantity 10 mg
Grade Analytical Standard