missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100 calcium binding protein A14 Antibody (OTI2G9) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73966
This item is not returnable.
View return policy
Description
S100 calcium binding protein A14 Monoclonal specifically detects S100 calcium binding protein A14 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| S100 calcium binding protein A14 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114 | |
| Mouse | |
| 11.5 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI2G9 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| S100A14 | |
| Full length human recombinant protein of human S100A14 (NP_065723) produced in E.coli. | |
| Affinity Purified | |
| Cancer | |
| 57402 | |
| Human, Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction