missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100 calcium binding protein A14 Antibody (S100A14/7403), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20352-20ug
This item is not returnable.
View return policy
Description
S100 calcium binding protein A14 Monoclonal antibody specifically detects S100 calcium binding protein A14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| S100 calcium binding protein A14 | |
| Monoclonal | |
| 0.2 mg/ml | |
| ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114 | |
| Recombinant full-length human S100 calcium binding protein A14 protein | |
| 20 μg | |
| Cancer | |
| 57402 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG2b κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| S100A14/7403 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction