missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115860
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total S100B.
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
Specifications
| S100 | |
| Polyclonal | |
| Unconjugated | |
| S100A1 | |
| AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20193, 20203, 25742, 295214, 6271, 6285 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P04271, P04631, P23297, P35467, P50114, P56565 | |
| S100A1, S100B | |
| A synthesized peptide derived from human S100B(Accession P04271), corresponding to amino acid residues D62-E92. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction