missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Monoclonal Antibody (OTI2A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807374S
This item is not returnable.
View return policy
Description
S100A1 Monoclonal antibody specifically detects S100A1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| S100A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52945 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3651 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A4 | |
| Unconjugated | |
| PDX1 | |
| AI266795; Protein S100-A1; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium-binding protein A1; S-100 protein alpha chain; S-100 protein subunit alpha; S100 protein, alpha polypeptide; S100a; S100A1; S100-alpha | |
| Full length human recombinant protein of human PDX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction