missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Recombinant Monoclonal [Clone S100A1/1706R] Rabbit, Clone: S100A1/1706R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 6271-RBM6-P0
This item is not returnable.
View return policy
Specifications
| S100A1 Recombinant Monoclonal [Clone S100A1/1706R] | |
| Cytoplasmic | |
| S100A1/1706R | |
| Unconjugated | |
| S100A1 | |
| Recombinant full-length human S100A1 protein | |
| Brain, Melanoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| Rabbit | |
| 10-12kDa | |
| Protein A/G Purified | |
| RUO | |
| 6271 | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction