Learn More
S100A10 Antibody (344015), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB2377MFV610
Product Code. 18582099
  • € 695.00 / 0.10mL
Description
S100A10 Monoclonal antibody specifically detects S100A10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S100A10
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Rat
0.1 mL
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
6281
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
344015
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11))
E. coli-derived recombinant mouse S100A10, Met1-Lys97, Accession # P08207