missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99718CL1
This item is not returnable.
View return policy
Description
S100A11 Polyclonal antibody specifically detects S100A11 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| S100A11 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) | |
| Produced in rabbits immunized with purified, recombinant Human S100A11 (Accession#: NP_005611.1; Met1-Thr105) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6282 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction