missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Mouse anti-Human, Clone: 20D11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59472-100UL
This item is not returnable.
View return policy
Description
S100A11 Monoclonal specifically detects S100A11 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| S100A11 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:10-1:1072, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 20D11 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| S100A11 | |
| Recombinant human S100A11 (1-105aa) purified from E. coli | |
| 100 μL | |
| Cell Cycle and Replication | |
| 6282 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only