Learn More
S100A11 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10237-100UL
Product Code. 17521526
  • € 450.00 / 100µL
Description
The S100A11 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human S100A11. This antibody recognizes human, rat, mouse antigen. The S100A11 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
S100A11
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
6282