missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ S100A11 Recombinant Rabbit Monoclonal Antibody (8U1Y8)
GREENER_CHOICE

Product Code. 18076241
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076241 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18076241 Supplier Invitrogen™ Supplier No. MA551258

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MAKISSPTET ERCIESLIAV FQKYAGKDGY NYTLSKTEFL SFMNTELAAF TKNQKDPGVL DRMMKKLDTN SDGQLDFSEF LNLIGGLAMA CHDSFLKAVP SQKRT.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8U1Y8
Concentration 0.076 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene S100A11
Gene Accession No. P31949
Gene Alias cal; Calgizzarin; calizzarin; EMAP; Emap1; EMAPI; endothelial monocyte activating polypeptide 1; endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; HEL-S-43; Metastatic lymph node gene 70 protein; MLN 70; MLN70; Protein S100-A11; Protein S100-A11, N-terminally processed; Protein S100C; Protein S100-C; S100 calcium binding protein A11; S100 calcium binding protein A11 (calgizzarin); S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; S100 calcium-binding protein A11 (calgizzarin); S100a11; S100a14; S100C
Gene Symbols S100A11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human S100A11 (NP_005611.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6282
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.