missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A13 Antibody (184), Alexa Fluor™ 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89547AF350
This item is not returnable.
View return policy
Description
S100A13 Monoclonal antibody specifically detects S100A13 in Mouse samples. It is validated for ELISA
Specifications
| S100A13 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A13 (P97352; Met1-Lys98). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6284 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 184 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction