missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A13 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98238AF350
This item is not returnable.
View return policy
Description
S100A13 Polyclonal specifically detects S100A13 in Mouse samples. It is validated for ELISA.
Specifications
| S100A13 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Alexa Fluor 350 | |
| S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
| Produced in rabbits immunized with purified, recombinant S100A13 (P97352; Met1-Lys98). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6284 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction