missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A16 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18447JF585
This item is not returnable.
View return policy
Description
S100A16 Polyclonal antibody specifically detects S100A16 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| S100A16 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| Aging-associated gene 13 protein, aging-associated protein 13, DT1P1A7, MGC17528, protein S100-A16, Protein S100-F, S100 calcium binding protein A16, S100FS100 calcium-binding protein A16 | |
| Recognizes region between residues Between Between 53 and 103.. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 140576 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction