missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A16 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99717MFV610
This item is not returnable.
View return policy
Description
S100A16 Polyclonal antibody specifically detects S100A16 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| S100A16 | |
| Rabbit | |
| Primary |
| ELISA | |
| Antigen and protein A Affinity-purified | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction