missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S100A16 Antibody (S100A16/7411) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27050B
This item is not returnable.
View return policy
Description
S100A16 Monoclonal antibody specifically detects S100A16 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100A16 | |
| Monoclonal | |
| Biotin | |
| Aging-associated gene 13 protein, aging-associated protein 13, DT1P1A7, MGC17528, protein S100-A16, Protein S100-F, S100 calcium binding protein A16, S100FS100 calcium-binding protein A16 | |
| Recombinant fragment (around aa1-103) of human S100A16 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 140576 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| S100A16/7411 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction