missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561104
This item is not returnable.
View return policy
Description
Immunogen sequence: VIVLVENFYK YVSKYSLVKN KISKSSFREM LQKELNHMLS DTGNRKAADK LIQNLDANHD GRISFDEYWT LIGGITGPIA KLIHEQEQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
S100A16 (S100 Calcium Binding Protein A16) is a Protein Coding gene. Diseases associated with S100A16 include Robinow Syndrome, Autosomal Recessive and Robinow Syndrome. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity. Calcium-binding protein. Binds one calcium ion per monomer. Can promote differentiation of adipocytes (in vitro). Overexpression in preadipocytes increases their proliferation, enhances adipogenesis and reduces insulin-stimulated glucose uptake.
Specifications
| S100A16 | |
| Polyclonal | |
| Unconjugated | |
| S100a16 | |
| 2300002L21Rik; AAG13; aging-associated gene 13 protein; aging-associated protein 13; AI325039; AI663996; DT1P1A7; MGC17528; protein S100-A16; Protein S100F; Protein S100-F; S100 calcium binding protein A16; S100 calcium-binding protein A16; S100A16; S100F | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140576 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96FQ6 | |
| S100a16 | |
| Recombinant protein corresponding to Human S100A16. Recombinant protein control fragment (Product #RP-94530). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction