missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A3 Antibody (001), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90583FR
This item is not returnable.
View return policy
Description
S100A3 Monoclonal specifically detects S100A3 in Mouse samples. It is validated for ELISA.
Specifications
| S100A3 | |
| Monoclonal | |
| DyLight 680 | |
| Protein S-100E, S100 calcium binding protein A3, S100 calcium-binding protein A3S100Eprotein S100-A3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A3 (NP_035440.1; Met1-Gln101). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 1 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6274 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction